Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TK1
Entrez Gene ID
7083
Gene Name
thymidine kinase 1
Gene Aliases
TK2
Chromosome Location
Chr.17: 78174079 - 78187208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003258.4 NP_003249.3 2 - 3 310 78
XM_017024992.1 XP_016880481.1 2 - 3 314 78
GenBank mRNA AB451268.1 - 2 - 3 100 78
AB451395.1 - 2 - 3 100 78
AK314950.1 - 2 - 3 153 78
BC006484.1 - 2 - 3 140 78
BC007872.2 - 2 - 3 127 78
BC007986.2 - 2 - 3 136 78
BI754079.1 - 2 - 3 340 78
BT006941.1 - 2 - 3 100 78
K02581.1 - 2 - 3 157 78

View Full Product Details