The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000948.5 | NP_000939.1 | 1 - 2 | 552 | 80 |
NM_001163558.2 | NP_001157030.1 | 2 - 3 | 224 | 80 | |
GenBank mRNA | BC015850.1 | - | 1 - 2 | 555 | 80 |
GQ305133.1 | - | 2 - 3 | 224 | 80 | |
X54393.1 | - | 2 - 3 | 79 |