The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001992.1 | NP_001001992.1 | 2 - 3 | 264 | 149 |
NM_001032410.1 | NP_001027582.1 | 3 - 4 | 327 | 149 | |
NM_006447.2 | NP_006438.1 | 2 - 3 | 264 | 149 | |
XM_017028257.1 | XP_016883746.1 | 3 - 4 | 411 | 149 | |
XM_017028258.1 | XP_016883747.1 | 3 - 4 | 293 | 149 | |
XM_017028259.1 | XP_016883748.1 | 3 - 4 | 265 | 149 | |
XM_017028261.1 | XP_016883750.1 | 3 - 4 | 268 | 149 | |
GenBank mRNA | AF126736.1 | - | 2 - 3 | 190 | 149 |
AK025104.1 | - | 2 - 3 | 230 | 149 | |
AK025989.1 | - | 2 - 3 | 230 | 149 | |
AK222681.1 | - | 2 - 3 | 193 | 149 | |
AK222884.1 | - | 2 - 3 | 186 | 149 | |
AY333928.1 | - | 2 - 3 | 190 | 149 | |
BC030777.2 | - | 2 - 3 | 220 | 149 | |
CB960929.1 | - | 2 - 3 | 264 | 149 |