Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EVC
Entrez Gene ID
2121
Gene Name
EvC ciliary complex subunit 1
Gene Aliases
DWF-1, EVC1, EVCL
Chromosome Location
Chr.4: 5711197 - 5829043 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306090.1 NP_001293019.1 6 - 7 990 114
NM_001306092.1 NP_001293021.1 6 - 7 990 114
NM_153717.2 NP_714928.1 6 - 7 990 114
XM_006713865.3 XP_006713928.1 6 - 7 987 114
XM_006713866.3 XP_006713929.1 6 - 7 987 114
XM_011513419.2 XP_011511721.1 6 - 7 984 114
XM_017007883.1 XP_016863372.1 6 - 7 984 114
GenBank mRNA AF216184.1 - 6 - 7 990 114
AF216185.1 - 6 - 7 990 114
AF239742.1 - 6 - 7 806 114
AK025394.1 - 1 - 2 119 114
BC085608.1 - 6 - 7 972 114

View Full Product Details