The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304548.1 | NP_001291477.1 | 8 - 9 | 1477 | 140 |
NM_152632.3 | NP_689845.2 | 8 - 9 | 1477 | 140 | |
XM_017029452.1 | XP_016884941.1 | 8 - 9 | 1470 | 140 | |
XM_017029453.1 | XP_016884942.1 | 8 - 9 | 1470 | 140 | |
GenBank mRNA | AK126295.1 | - | 8 - 9 | 1443 | 140 |
BC027936.1 | - | 8 - 9 | 1430 | 140 |