The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019070.4 | NP_061943.2 | 4 - 5 | 516 | 67 |
NR_033677.1 | 4 - 5 | 445 | 67 | ||
XM_011528083.1 | XP_011526385.1 | 5 - 6 | 564 | 67 | |
XM_011528084.2 | XP_011526386.1 | 5 - 6 | 508 | 67 | |
GenBank mRNA | AK001294.1 | - | 4 - 5 | 465 | 67 |
AK223294.1 | - | 4 - 5 | 459 | 67 | |
AK291916.1 | - | 4 - 5 | 476 | 67 | |
BC000979.3 | - | 5 - 6 | 540 | 67 | |
BC002674.2 | - | 4 - 5 | 506 | 67 | |
BP252678.1 | - | 4 - 5 | 419 | 67 | |
BX647408.1 | - | 5 - 6 | 383 | 67 | |
DA136213.1 | - | 4 - 5 | 516 | 67 |