The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019070.4 | NP_061943.2 | 8 - 9 | 995 | 92 |
NR_033677.1 | 8 - 9 | 924 | 92 | ||
XM_011528083.1 | XP_011526385.1 | 9 - 10 | 1043 | 92 | |
XM_011528084.2 | XP_011526386.1 | 9 - 10 | 987 | 92 | |
GenBank mRNA | AK001294.1 | - | 8 - 9 | 943 | 92 |
AK223294.1 | - | 8 - 9 | 938 | 92 | |
AK291916.1 | - | 8 - 9 | 955 | 92 | |
BC002674.2 | - | 8 - 9 | 985 | 92 | |
BX647408.1 | - | 9 - 10 | 862 | 92 |