The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019070.4 | NP_061943.2 | 2 - 3 | 308 | 59 |
NR_033677.1 | 2 - 3 | 237 | 59 | ||
GenBank mRNA | AK223294.1 | - | 2 - 3 | 251 | 59 |
AK291916.1 | - | 2 - 3 | 268 | 59 | |
BC002674.2 | - | 2 - 3 | 298 | 59 | |
BP252678.1 | - | 2 - 3 | 211 | 59 | |
BX647408.1 | - | 3 - 4 | 175 | 59 |