The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020665.5 | NP_065716.1 | 2 - 3 | 373 | 142 |
XM_017029681.1 | XP_016885170.1 | 2 - 3 | 648 | 142 | |
GenBank mRNA | AF229179.1 | - | 2 - 3 | 140 | 142 |
AK313835.1 | - | 2 - 3 | 198 | 142 | |
AY359060.1 | - | 2 - 3 | 123 | 142 | |
BC014317.1 | - | 2 - 3 | 377 | 142 | |
BC015099.1 | - | 2 - 3 | 205 | 142 | |
BC050606.1 | - | 2 - 3 | 125 | 142 |