The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193416.2 | NP_001180345.1 | 6 - 7 | 635 | 83 |
NM_001193417.2 | NP_001180346.1 | 5 - 6 | 587 | 83 | |
NM_001356.4 | NP_001347.3 | 6 - 7 | 1489 | 83 | |
NR_126093.1 | 6 - 7 | 1489 | 83 | ||
XM_011543892.2 | XP_011542194.1 | 6 - 7 | 666 | 83 | |
XM_017029313.1 | XP_016884802.1 | 6 - 7 | 2834 | 83 | |
XM_017029314.1 | XP_016884803.1 | 6 - 7 | 2834 | 83 | |
GenBank mRNA | AB208983.1 | - | 6 - 7 | 587 | 83 |
AB451220.1 | - | 6 - 7 | 544 | 83 | |
AB451343.1 | - | 6 - 7 | 544 | 83 | |
AF000982.1 | - | 6 - 7 | 1400 | 83 | |
AF000983.1 | - | 6 - 7 | 1400 | 83 | |
AF061337.1 | - | 6 - 7 | 619 | 83 | |
AK291153.1 | - | 6 - 7 | 635 | 83 | |
AK297159.1 | - | 6 - 7 | 635 | 83 | |
AK304661.1 | - | 6 - 7 | 678 | 83 | |
AK304689.1 | - | 5 - 6 | 587 | 83 | |
AK310891.1 | - | 5 - 6 | 587 | 83 | |
BC011819.2 | - | 6 - 7 | 614 | 83 | |
U50553.1 | - | 6 - 7 | 853 | 83 |