Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRC5
Entrez Gene ID
84166
Gene Name
NLR family CARD domain containing 5
Gene Aliases
CLR16.1, NOD27, NOD4
Chromosome Location
Chr.16: 56989547 - 57083524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032206.4 NP_115582.4 27 - 28 3951 72
XM_005256193.2 XP_005256250.1 28 - 29 4012 72
XM_005256194.3 XP_005256251.1 28 - 29 4223 72
XM_005256195.2 XP_005256252.1 28 - 29 4009 72
XM_005256196.2 XP_005256253.1 27 - 28 3928 72
XM_005256197.2 XP_005256254.1 27 - 28 3925 72
XM_005256199.3 XP_005256256.1 28 - 29 4009 72
XM_005256201.3 XP_005256258.1 27 - 28 3919 72
XM_006721297.2 XP_006721360.1 28 - 29 4000 72
XM_006721298.2 XP_006721361.1 27 - 28 3922 72
XM_006721300.2 XP_006721363.1 28 - 29 4012 72
XM_011523373.2 XP_011521675.1 28 - 29 4159 72
XM_011523375.2 XP_011521677.1 28 - 29 4012 72
XM_011523376.2 XP_011521678.1 23 - 24 1838 72
XM_017023766.1 XP_016879255.1 28 - 29 4012 72
XM_017023767.1 XP_016879256.1 27 - 28 3913 72
XM_017023768.1 XP_016879257.1 28 - 29 4009 72
XM_017023769.1 XP_016879258.1 28 - 29 4000 72
XM_017023770.1 XP_016879259.1 28 - 29 4012 72
XM_017023771.1 XP_016879260.1 28 - 29 4009 72
GenBank mRNA AF389420.1 - 26 - 27 3824 72
AK025362.1 - 23 - 24 3080 72
AK027414.1 - 7 - 8 541 72
AK027416.1 - 7 - 8 540 72
AK074133.1 - 16 - 17 1277 72
AK090439.1 - 23 - 24 3006 72
BC050527.1 - 11 - 12 829 72
BC063566.1 - 10 - 11 737 72
BX640842.1 - 23 - 24 2393 72
EF452236.1 - 26 - 27 3824 72

View Full Product Details