The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173833.5 | NP_776194.2 | 8 - 9 | 1964 | 57 |
GenBank mRNA | AK172746.1 | - | 8 - 9 | 1794 | 57 |
AK301006.1 | - | 6 - 7 | 605 | 57 | |
AY358150.1 | - | 7 - 8 | 1112 | 57 |