The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207034.2 | NP_996917.1 | 4 - 5 | 980 | 81 |
XM_005260313.4 | XP_005260370.1 | 4 - 5 | 1006 | 81 | |
GenBank mRNA | BC008876.2 | - | 4 - 5 | 980 | 81 |
BT007085.1 | - | 4 - 5 | 594 | 81 | |
J05081.1 | - | 4 - 5 | 787 | 81 |