The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172766.2 | NP_001166237.1 | 15 - 16 | 2209 | 80 |
NM_014491.3 | NP_055306.1 | 15 - 16 | 2212 | 80 | |
NM_148898.3 | NP_683696.2 | 16 - 17 | 2287 | 80 | |
NM_148900.3 | NP_683698.2 | 16 - 17 | 2263 | 80 | |
NR_033766.1 | 18 - 19 | 2223 | 80 | ||
NR_033767.1 | 16 - 17 | 2270 | 80 | ||
XM_017012801.1 | XP_016868290.1 | 18 - 19 | 2893 | 80 | |
GenBank mRNA | AF337817.1 | - | 15 - 16 | 1988 | 80 |
AK131266.1 | - | 18 - 19 | 2207 | 80 | |
AK296957.1 | - | 16 - 17 | 2143 | 80 | |
AY144615.1 | - | 16 - 17 | 2282 | 80 | |
BC126104.1 | - | 16 - 17 | 1944 | 80 | |
BC143866.1 | - | 16 - 17 | 1944 | 80 | |
BC143867.1 | - | 15 - 16 | 1883 | 80 | |
CR749236.1 | - | 22 - 23 | 2593 | 80 | |
DQ778626.1 | - | 10 - 11 | 1283 | 80 | |
KJ534839.1 | - | 15 - 16 | 1913 | 80 | |
KJ535079.1 | - | 14 - 15 | 2051 | 80 | |
KJ535086.1 | - | 14 - 15 | 1837 | 80 |