Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NALCN
Entrez Gene ID
259232
Gene Name
sodium leak channel, non-selective
Gene Aliases
CLIFAHDD, CanIon, IHPRF, IHPRF1, INNFD, VGCNL1, bA430M15.1
Chromosome Location
Chr.13: 101053774 - 101417206 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052867.2 NP_443099.1 31 - 32 112
XM_011521067.2 XP_011519369.1 31 - 32 112
XM_011521068.2 XP_011519370.1 31 - 32 112
XM_011521069.2 XP_011519371.1 30 - 31 112
XM_017020534.1 XP_016876023.1 30 - 31 112
XM_017020535.1 XP_016876024.1 30 - 31 112
XM_017020536.1 XP_016876025.1 26 - 27 112
XM_017020537.1 XP_016876026.1 25 - 26 112
GenBank mRNA AK126621.1 - 12 - 13 112
AK126718.1 - 4 - 5 112
AY141972.1 - 31 - 32 3665 112
DA128114.1 - 2 - 3 112

View Full Product Details