The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052867.2 | NP_443099.1 | 3 - 4 | 478 | 77 |
XM_011521067.2 | XP_011519369.1 | 3 - 4 | 418 | 77 | |
XM_011521068.2 | XP_011519370.1 | 3 - 4 | 401 | 77 | |
XM_011521069.2 | XP_011519371.1 | 3 - 4 | 418 | 77 | |
XM_017020534.1 | XP_016876023.1 | 3 - 4 | 449 | 77 | |
XM_017020535.1 | XP_016876024.1 | 3 - 4 | 391 | 77 | |
GenBank mRNA | AK094669.1 | - | 3 - 4 | 449 | 77 |
AY141972.1 | - | 3 - 4 | 372 | 77 | |
BC028390.1 | - | 3 - 4 | 385 | 77 | |
BC064343.1 | - | 2 - 3 | 317 | 77 |