The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318202.1 | NP_001305131.1 | 19 - 20 | 2854 | 84 |
NM_013241.2 | NP_037373.2 | 17 - 18 | 2776 | 84 | |
XM_006721180.1 | XP_006721243.1 | 8 - 9 | 1429 | 84 | |
XM_011523043.2 | XP_011521345.1 | 19 - 20 | 3070 | 84 | |
XM_011523045.2 | XP_011521347.1 | 19 - 20 | 2564 | 84 | |
GenBank mRNA | AB041046.1 | - | 17 - 18 | 2746 | 84 |
AB209584.1 | - | 15 - 16 | 3245 | 84 | |
AF113615.1 | - | 17 - 18 | 2682 | 84 | |
AK025992.1 | - | 3 - 4 | 466 | 84 | |
AK301157.1 | - | 7 - 8 | 1487 | 84 | |
AY192154.1 | - | 17 - 18 | 2664 | 84 | |
BC033084.1 | - | 17 - 18 | 2721 | 84 | |
KJ658417.1 | - | 19 - 20 | 2742 | 84 |