The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322194.1 | NP_001309123.1 | 22 - 23 | 3657 | 116 |
NM_001322195.1 | NP_001309124.1 | 21 - 22 | 3175 | 116 | |
NM_001322196.1 | NP_001309125.1 | 21 - 22 | 3488 | 116 | |
NM_001322198.1 | NP_001309127.1 | 22 - 23 | 3476 | 116 | |
NM_001322199.1 | NP_001309128.1 | 22 - 23 | 3418 | 116 | |
NM_001322204.1 | NP_001309133.1 | 19 - 20 | 2927 | 116 | |
NM_004972.3 | NP_004963.1 | 22 - 23 | 3556 | 116 | |
GenBank mRNA | AF001362.1 | - | 20 - 21 | 3062 | 116 |
AF005216.1 | - | 22 - 23 | 3556 | 116 | |
AF058925.1 | - | 22 - 23 | 3539 | 116 | |
AK292525.1 | - | 21 - 22 | 3175 | 116 | |
AK302618.1 | - | 19 - 20 | 2986 | 116 | |
AY973034.1 | - | 21 - 22 | 3133 | 116 | |
BC039695.1 | - | 21 - 22 | 3160 | 116 |