The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152263.3 | NP_689476.2 | 2 - 3 | 363 | 73 |
GenBank mRNA | BC008407.1 | - | 2 - 3 | 329 | 73 |
BC008425.1 | - | 2 - 3 | 332 | 73 | |
DA900740.1 | - | 2 - 3 | 364 | 73 | |
X04201.1 | - | 2 - 3 | 304 | 73 |