Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
R3HDM2
Entrez Gene ID
22864
Gene Name
R3H domain containing 2
Gene Aliases
CAG6, PR01365
Chromosome Location
Chr.12: 57253764 - 57430999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014925.3 NP_055740.3 18 - 19 2299 61
XM_005268725.3 XP_005268782.1 20 - 21 2649 61
XM_011538031.1 XP_011536333.1 21 - 22 2537 61
XM_011538033.1 XP_011536335.1 22 - 23 2591 61
XM_011538035.1 XP_011536337.1 21 - 22 2495 61
XM_011538036.1 XP_011536338.1 21 - 22 2489 61
XM_011538037.1 XP_011536339.1 20 - 21 2471 61
XM_011538038.1 XP_011536340.1 20 - 21 2441 61
XM_011538039.1 XP_011536341.1 20 - 21 2435 61
XM_011538040.1 XP_011536342.1 21 - 22 2489 61
XM_011538042.2 XP_011536344.1 23 - 24 2687 61
XM_017018998.1 XP_016874487.1 20 - 21 2513 61
XM_017018999.1 XP_016874488.1 23 - 24 2728 61
XM_017019000.1 XP_016874489.1 21 - 22 2495 61
XM_017019001.1 XP_016874490.1 22 - 23 2801 61
XM_017019002.1 XP_016874491.1 20 - 21 2465 61
XM_017019003.1 XP_016874492.1 20 - 21 2441 61
XM_017019004.1 XP_016874493.1 20 - 21 2435 61
XM_017019005.1 XP_016874494.1 22 - 23 2646 61
XM_017019006.1 XP_016874495.1 21 - 22 2539 61
XM_017019007.1 XP_016874496.1 21 - 22 2574 61
XM_017019008.1 XP_016874497.1 19 - 20 2339 61
XM_017019009.1 XP_016874498.1 20 - 21 2393 61
XM_017019010.1 XP_016874499.1 20 - 21 2467 61
XM_017019011.1 XP_016874500.1 23 - 24 2690 61
XM_017019012.1 XP_016874501.1 19 - 20 2375 61
XM_017019013.1 XP_016874502.1 20 - 21 2509 61
XM_017019014.1 XP_016874503.1 20 - 21 2544 61
XM_017019015.1 XP_016874504.1 20 - 21 2686 61
XM_017019016.1 XP_016874505.1 21 - 22 2705 61
XM_017019017.1 XP_016874506.1 20 - 21 2467 61
XM_017019018.1 XP_016874507.1 20 - 21 2433 61
XM_017019019.1 XP_016874508.1 21 - 22 2529 61
XM_017019020.1 XP_016874509.1 18 - 19 2279 61
XM_017019021.1 XP_016874510.1 20 - 21 2503 61
XM_017019022.1 XP_016874511.1 19 - 20 2413 61
XM_017019023.1 XP_016874512.1 19 - 20 2424 61
XM_017019024.1 XP_016874513.1 19 - 20 2413 61
XM_017019025.1 XP_016874514.1 19 - 20 2378 61
XM_017019026.1 XP_016874515.1 20 - 21 2418 61
XM_017019027.1 XP_016874516.1 19 - 20 2549 61
XM_017019028.1 XP_016874517.1 18 - 19 2311 61
XM_017019029.1 XP_016874518.1 18 - 19 2397 61
XM_017019030.1 XP_016874519.1 11 - 12 1678 61
XM_017019031.1 XP_016874520.1 14 - 15 1962 61
XM_017019032.1 XP_016874521.1 10 - 11 1530 61
XM_017019033.1 XP_016874522.1 12 - 13 1681 61
GenBank mRNA AB023219.1 - 20 - 21 2656 61
AK293394.1 - 10 - 11 1551 61
AX775761.1 - 20 - 21 2656 61
AX775763.1 - 20 - 21 2698 61
BC041857.1 - 10 - 11 2111 61
BC104995.1 - 8 - 9 1306 61
BC112226.1 - 8 - 9 1306 61
BC143605.1 - 9 - 10 1408 61

View Full Product Details