The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105513.2 | NP_001098983.2 | 9 - 10 | 1250 | 71 |
NM_145755.2 | NP_665698.2 | 10 - 11 | 1397 | 71 | |
XM_005264921.4 | XP_005264978.1 | 10 - 11 | 1323 | 71 | |
XM_005264922.4 | XP_005264979.1 | 10 - 11 | 1323 | 71 | |
XM_005264923.3 | XP_005264980.1 | 10 - 11 | 1323 | 71 | |
XM_005264924.4 | XP_005264981.1 | 10 - 11 | 1323 | 71 | |
XM_005264925.4 | XP_005264982.1 | 9 - 10 | 1200 | 71 | |
XM_005264926.4 | XP_005264983.1 | 10 - 11 | 1323 | 71 | |
XM_005264927.4 | XP_005264984.1 | 6 - 7 | 777 | 71 | |
XM_006713011.3 | XP_006713074.1 | 10 - 11 | 1323 | 71 | |
XM_011533447.2 | XP_011531749.1 | 9 - 10 | 1224 | 71 | |
XM_011533449.1 | XP_011531751.1 | 10 - 11 | 1323 | 71 | |
XM_017005843.1 | XP_016861332.1 | 10 - 11 | 1323 | 71 | |
GenBank mRNA | AJ487015.1 | - | 10 - 11 | 1359 | 71 |
AK302408.1 | - | - | 1233 | 71 | |
AK302465.1 | - | 9 - 10 | 1104 | 71 | |
BC129948.1 | - | 10 - 11 | 1305 | 71 |