The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006080.2 | NP_006071.1 | 1 - 2 | 433 | 71 |
XM_005250110.3 | XP_005250167.1 | 4 - 5 | 628 | 71 | |
XM_005250111.4 | XP_005250168.1 | 3 - 4 | 468 | 71 | |
XM_006715839.3 | XP_006715902.1 | 2 - 3 | 371 | 71 | |
XM_011515734.2 | XP_011514036.1 | 4 - 5 | 453 | 71 | |
XM_017011673.1 | XP_016867162.1 | 2 - 3 | 288 | 71 | |
GenBank mRNA | AK289954.1 | - | 1 - 2 | 526 | 71 |
BC111416.1 | - | 1 - 2 | 132 | 71 | |
BX491494.1 | - | 1 - 2 | 470 | 71 | |
DR002146.1 | - | 1 - 2 | 271 | 71 | |
L26081.1 | - | 1 - 2 | 133 | 71 |