The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006080.2 | NP_006071.1 | 5 - 6 | 859 | 73 |
XM_005250110.3 | XP_005250167.1 | 8 - 9 | 1054 | 73 | |
XM_005250111.4 | XP_005250168.1 | 7 - 8 | 894 | 73 | |
XM_006715839.3 | XP_006715902.1 | 6 - 7 | 797 | 73 | |
XM_011515734.2 | XP_011514036.1 | 8 - 9 | 879 | 73 | |
XM_017011673.1 | XP_016867162.1 | 6 - 7 | 714 | 73 | |
GenBank mRNA | AK289954.1 | - | 5 - 6 | 952 | 73 |
BC111416.1 | - | 5 - 6 | 558 | 73 | |
DR002146.1 | - | 5 - 6 | 697 | 73 | |
L26081.1 | - | 5 - 6 | 559 | 73 |