Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGO3
Entrez Gene ID
192669
Gene Name
argonaute 3, RISC catalytic component
Gene Aliases
EIF2C3
Chromosome Location
Chr.1: 35925198 - 36072037 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024852.3 NP_079128.2 3 - 4 653 65
XM_005270575.3 XP_005270632.1 4 - 5 6107 65
XM_011540879.2 XP_011539181.1 3 - 4 1535 65
XM_017000523.1 XP_016856012.1 4 - 5 6123 65
GenBank mRNA AK022827.1 - 3 - 4 408 65
AK027796.1 - 4 - 5 587 65
AK292870.1 - 3 - 4 619 65
BC066888.1 - 3 - 4 653 65

View Full Product Details