The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016184.3 | NP_057268.1 | 1 - 2 | 327 | 110 |
NM_194450.2 | NP_919432.1 | 1 - 2 | 327 | 110 | |
GenBank mRNA | AF067800.1 | - | 1 - 2 | 348 | 110 |
AF109146.1 | - | 1 - 2 | 174 | 110 | |
AF200738.1 | - | 1 - 2 | 80 | 110 | |
AF328684.1 | - | 1 - 2 | 319 | 110 | |
AJ133532.1 | - | 1 - 2 | 321 | 110 | |
BC074822.2 | - | 1 - 2 | 102 | 110 | |
BC074874.2 | - | 1 - 2 | 102 | 110 | |
BC117439.1 | - | 1 - 2 | 222 | 110 | |
BC117441.1 | - | 1 - 2 | 222 | 110 |