The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171201.1 | NP_001164672.1 | 4 - 5 | 1465 | 71 |
NM_001171202.1 | NP_001164673.1 | 4 - 5 | 1381 | 71 | |
NM_001171203.2 | NP_001164674.1 | 6 - 7 | 1661 | 71 | |
NM_001171204.2 | NP_001164675.1 | 6 - 7 | 1629 | 71 | |
NM_016525.4 | NP_057609.2 | 5 - 6 | 1506 | 71 | |
NR_033243.2 | 6 - 7 | 1622 | 71 | ||
XM_006716779.3 | XP_006716842.1 | 5 - 6 | 1532 | 71 | |
XM_011517898.2 | XP_011516200.1 | 4 - 5 | 1299 | 71 | |
XM_011517899.1 | XP_011516201.1 | 6 - 7 | 1563 | 71 | |
XM_011517900.1 | XP_011516202.1 | 5 - 6 | 1438 | 71 | |
XM_017014801.1 | XP_016870290.1 | 6 - 7 | 1898 | 71 | |
XM_017014802.1 | XP_016870291.1 | 4 - 5 | 1255 | 71 | |
GenBank mRNA | AF222043.2 | - | 5 - 6 | 1435 | 71 |
AK074426.1 | - | 5 - 6 | 1499 | 71 | |
AK074724.1 | - | 3 - 4 | 1244 | 71 | |
AK074745.1 | - | 5 - 6 | 1466 | 71 | |
AK074812.1 | - | 5 - 6 | 1506 | 71 | |
AK074969.1 | - | 6 - 7 | 1608 | 71 | |
AK074995.1 | - | 5 - 6 | 1470 | 71 | |
AK223235.1 | - | 5 - 6 | 1461 | 71 | |
AK295482.1 | - | 4 - 5 | 1341 | 71 | |
AK303711.1 | - | 4 - 5 | 1465 | 71 | |
AK309329.1 | - | 4 - 5 | 1336 | 71 | |
AL136733.1 | - | 5 - 6 | 1299 | 71 | |
BC020950.1 | - | 5 - 6 | 1468 | 71 | |
BC098141.1 | - | 5 - 6 | 1402 | 71 | |
BC098316.1 | - | 5 - 6 | 1402 | 71 | |
BC099726.1 | - | 5 - 6 | 1402 | 71 | |
BC100668.1 | - | 5 - 6 | 1402 | 71 | |
CR533551.1 | - | 4 - 5 | 1263 | 71 |