The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018439.3 | NP_060909.1 | 1 - 2 | 148 | 89 |
GenBank mRNA | AB026264.1 | - | 1 - 2 | 75 | 89 |
AK292533.1 | - | 1 - 2 | 148 | 89 | |
AK312727.1 | - | 1 - 2 | 123 | 89 | |
BC034016.1 | - | 1 - 2 | 96 | 89 |