Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VEPH1
Entrez Gene ID
79674
Gene Name
ventricular zone expressed PH domain containing 1
Gene Aliases
MELT, VEPH
Chromosome Location
Chr.3: 157259742 - 157503626 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167912.1 NP_001161384.1 11 - 12 2243 69
NM_024621.2 NP_078897.2 11 - 12 2434 69
XM_011513134.2 XP_011511436.1 10 - 11 2177 69
XM_011513135.1 XP_011511437.1 11 - 12 2073 69
GenBank mRNA AB051479.1 - 11 - 12 2243 69
AK022666.1 - 1 - 2 111 69
AK027625.1 - 11 - 12 2332 69
AL713656.1 - 10 - 11 2031 69
BC026235.1 - 3 - 4 556 69
BC042159.1 - 4 - 5 893 69
BC101660.1 - 10 - 11 2070 69
BC113555.1 - 10 - 11 2070 69

View Full Product Details