Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VEPH1
Entrez Gene ID
79674
Gene Name
ventricular zone expressed PH domain containing 1
Gene Aliases
MELT, VEPH
Chromosome Location
Chr.3: 157259742 - 157503626 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167911.1 NP_001161383.1 11 - 12 2220 68
NM_001167912.1 NP_001161384.1 12 - 13 2355 68
NM_024621.2 NP_078897.2 12 - 13 2546 68
XM_011513134.2 XP_011511436.1 11 - 12 2289 68
XM_011513135.1 XP_011511437.1 12 - 13 2185 68
GenBank mRNA AB051479.1 - 12 - 13 2355 68
AK022666.1 - 2 - 3 223 68
AK027625.1 - 12 - 13 2444 68
AL713656.1 - 11 - 12 2143 68
BC026235.1 - 4 - 5 668 68
BC042159.1 - 5 - 6 1005 68
BC101660.1 - 11 - 12 2182 68
BC111017.1 - 11 - 12 2212 68
BC113555.1 - 11 - 12 2182 68

View Full Product Details