The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321190.1 | NP_001308119.1 | 5 - 6 | 929 | 63 |
NM_001321191.1 | NP_001308120.1 | 5 - 6 | 838 | 63 | |
NM_006666.2 | NP_006657.1 | 5 - 6 | 859 | 63 | |
NR_135578.1 | 5 - 6 | 859 | 63 | ||
XM_011526330.1 | XP_011524632.1 | 5 - 6 | 772 | 63 | |
GenBank mRNA | AB024301.1 | - | 5 - 6 | 408 | 63 |
AF124607.1 | - | 5 - 6 | 416 | 63 | |
AF151804.1 | - | 5 - 6 | 401 | 63 | |
AF155138.1 | - | 5 - 6 | 401 | 63 | |
AK027762.1 | - | 5 - 6 | 479 | 63 | |
AK057498.1 | - | 5 - 6 | 1406 | 63 | |
AK074542.1 | - | 5 - 6 | 409 | 63 | |
AK301344.1 | - | 5 - 6 | 417 | 63 | |
AL136743.1 | - | 4 - 5 | 398 | 63 | |
BC000428.2 | - | 5 - 6 | 400 | 63 | |
BC004531.2 | - | 5 - 6 | 408 | 63 | |
BC008355.1 | - | 5 - 6 | 408 | 63 | |
CR533507.1 | - | 4 - 5 | 395 | 63 | |
Y18417.1 | - | 5 - 6 | 400 | 63 |