The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000203.4 | NP_000194.2 | 3 - 4 | 470 | 95 |
NR_110313.1 | 3 - 4 | 470 | 95 | ||
XM_011513461.2 | XP_011511763.1 | 2 - 3 | 615 | 95 | |
XM_017008163.1 | XP_016863652.1 | 3 - 4 | 418 | 95 | |
GenBank mRNA | AK098805.1 | - | 2 - 3 | 438 | 95 |
AK125223.1 | - | 2 - 3 | 282 | 95 | |
AK291816.1 | - | 3 - 4 | 403 | 95 | |
BC029959.1 | - | 3 - 4 | 394 | 95 | |
DA057148.1 | - | 3 - 4 | 418 | 95 | |
M74715.1 | - | 3 - 4 | 470 | 95 |