The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000203.4 | NP_000194.2 | 7 - 8 | 1059 | 155 |
NR_110313.1 | 7 - 8 | 1059 | 155 | ||
XM_011513461.2 | XP_011511763.1 | 6 - 7 | 1204 | 155 | |
XM_017008163.1 | XP_016863652.1 | 6 - 7 | 1513 | 155 | |
GenBank mRNA | AK098805.1 | - | 6 - 7 | 1027 | 155 |
AK291816.1 | - | 7 - 8 | 992 | 155 | |
BC029959.1 | - | 7 - 8 | 983 | 155 | |
M74715.1 | - | 7 - 8 | 1059 | 155 |