Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CENPV
Entrez Gene ID
201161
Gene Name
centromere protein V
Gene Aliases
3110013H01Rik, CENP-V, PRR6, p30
Chromosome Location
Chr.17: 16342534 - 16353498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181716.2 NP_859067.2 4 - 5 753 79
GenBank mRNA AF514992.1 - 4 - 5 689 79
AY382899.1 - 2 - 3 727 79
BC052604.1 - 4 - 5 763 79
BC137486.1 - 4 - 5 785 79
BC137487.1 - 4 - 5 785 79
BU618545.1 - 1 - 2 79

View Full Product Details