The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164883.1 | NP_001158355.1 | 15 - 16 | 2464 | 97 |
NM_001282597.2 | NP_001269526.1 | 15 - 16 | 2464 | 97 | |
NM_001282598.1 | NP_001269527.1 | 15 - 16 | 2317 | 97 | |
NM_001282599.1 | NP_001269528.1 | 9 - 10 | 1405 | 97 | |
NM_001282600.1 | NP_001269529.1 | 9 - 10 | 1348 | 97 | |
NM_001320810.1 | NP_001307739.1 | 9 - 10 | 1348 | 97 | |
NM_004389.3 | NP_004380.2 | 15 - 16 | 2464 | 97 | |
XM_011532555.2 | XP_011530857.1 | 15 - 16 | 2311 | 97 | |
XM_011532556.2 | XP_011530858.1 | 16 - 17 | 2507 | 97 | |
XM_011532557.2 | XP_011530859.1 | 7 - 8 | 1053 | 97 | |
XM_017003403.1 | XP_016858892.1 | 16 - 17 | 2507 | 97 | |
XM_017003404.1 | XP_016858893.1 | 15 - 16 | 2402 | 97 | |
XM_017003405.1 | XP_016858894.1 | 15 - 16 | 2402 | 97 | |
XM_017003406.1 | XP_016858895.1 | 9 - 10 | 1438 | 97 | |
GenBank mRNA | AK127226.1 | - | 19 - 20 | 2910 | 97 |
AK295181.1 | - | 15 - 16 | 2263 | 97 | |
AK295493.1 | - | 9 - 10 | 1335 | 97 | |
AK303035.1 | - | 15 - 16 | 2317 | 97 | |
BC040458.1 | - | 14 - 15 | 2136 | 97 | |
BC052996.1 | - | 15 - 16 | 2398 | 97 | |
BX537769.1 | - | 9 - 10 | 1405 | 97 | |
BX648368.1 | - | 2 - 3 | 277 | 97 | |
HQ589336.1 | - | 9 - 10 | 1336 | 97 | |
M94151.2 | - | 14 - 15 | 2215 | 97 |