The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000352.4 | NP_000343.2 | 12 - 13 | 1885 | 64 |
NM_001287174.1 | NP_001274103.1 | 12 - 13 | 1885 | 64 | |
XM_011520331.2 | XP_011518633.1 | 12 - 13 | 1907 | 64 | |
XM_017018197.1 | XP_016873686.1 | 12 - 13 | 1879 | 64 | |
XM_017018198.1 | XP_016873687.1 | 12 - 13 | 1879 | 64 | |
XM_017018199.1 | XP_016873688.1 | 12 - 13 | 1884 | 64 | |
XM_017018200.1 | XP_016873689.1 | 12 - 13 | 1903 | 64 | |
XM_017018201.1 | XP_016873690.1 | 12 - 13 | 1879 | 64 | |
XM_017018202.1 | XP_016873691.1 | 11 - 12 | 1680 | 64 | |
XM_017018203.1 | XP_016873692.1 | 12 - 13 | 1879 | 64 | |
XM_017018204.1 | XP_016873693.1 | 6 - 7 | 861 | 64 | |
GenBank mRNA | AF087138.1 | - | 12 - 13 | 1851 | 64 |
L78207.1 | - | 12 - 13 | 1854 | 64 | |
R84665.1 | - | 3 - 4 | 208 | 64 | |
U63421.1 | - | 12 - 13 | 1816 | 64 |