The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000352.4 | NP_000343.2 | 15 - 16 | 2187 | 59 |
NM_001287174.1 | NP_001274103.1 | 15 - 16 | 2187 | 59 | |
XM_011520331.2 | XP_011518633.1 | 15 - 16 | 2209 | 59 | |
XM_017018197.1 | XP_016873686.1 | 15 - 16 | 2247 | 59 | |
XM_017018198.1 | XP_016873687.1 | 15 - 16 | 2247 | 59 | |
XM_017018199.1 | XP_016873688.1 | 15 - 16 | 2252 | 59 | |
XM_017018200.1 | XP_016873689.1 | 15 - 16 | 2205 | 59 | |
XM_017018201.1 | XP_016873690.1 | 15 - 16 | 2247 | 59 | |
XM_017018202.1 | XP_016873691.1 | 14 - 15 | 2048 | 59 | |
XM_017018203.1 | XP_016873692.1 | 15 - 16 | 2247 | 59 | |
XM_017018204.1 | XP_016873693.1 | 9 - 10 | 1163 | 59 | |
GenBank mRNA | AB209084.1 | - | 15 - 16 | 2131 | 59 |
AF087138.1 | - | 15 - 16 | 2153 | 59 | |
L78207.1 | - | 15 - 16 | 2156 | 59 | |
U63421.1 | - | 15 - 16 | 2118 | 59 |