The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014326.3 | NP_055141.2 | 6 - 7 | 663 | 92 |
XM_011521413.2 | XP_011519715.1 | 5 - 6 | 638 | 92 | |
XM_011521414.1 | XP_011519716.1 | 5 - 6 | 638 | 92 | |
XM_011521415.2 | XP_011519717.1 | 5 - 6 | 638 | 92 | |
XM_011521416.2 | XP_011519718.1 | 6 - 7 | 811 | 92 | |
XM_011521417.1 | XP_011519719.1 | 5 - 6 | 638 | 92 | |
XM_011521421.2 | XP_011519723.1 | 4 - 5 | 448 | 92 | |
XM_017022043.1 | XP_016877532.1 | 4 - 5 | 407 | 92 | |
XM_017022044.1 | XP_016877533.1 | 5 - 6 | 638 | 92 | |
XM_017022045.1 | XP_016877534.1 | 5 - 6 | 638 | 92 | |
XM_017022046.1 | XP_016877535.1 | 5 - 6 | 638 | 92 | |
XM_017022047.1 | XP_016877536.1 | 4 - 5 | 448 | 92 | |
XM_017022048.1 | XP_016877537.1 | 3 - 4 | 397 | 92 | |
XM_017022049.1 | XP_016877538.1 | 4 - 5 | 444 | 92 | |
XM_017022050.1 | XP_016877539.1 | 3 - 4 | 249 | 92 | |
XM_017022051.1 | XP_016877540.1 | 3 - 4 | 395 | 92 | |
GenBank mRNA | AB018001.1 | - | 6 - 7 | 662 | 92 |
AF052941.1 | - | 6 - 7 | 663 | 92 | |
BC114506.1 | - | 5 - 6 | 652 | 92 | |
BC114951.1 | - | 5 - 6 | 560 | 92 | |
GU056176.1 | - | 6 - 7 | 663 | 92 |