The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007038.4 | NP_008969.2 | 2 - 3 | 1964 | 91 |
GenBank mRNA | AF142099.1 | - | 2 - 3 | 1357 | 91 |
AK094057.1 | - | 2 - 3 | 1963 | 91 | |
BC093775.1 | - | 2 - 3 | 1319 | 91 | |
BC093777.1 | - | 2 - 3 | 1319 | 91 |