The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018161.4 | NP_060631.2 | 19 - 20 | 2025 | 86 |
GenBank mRNA | AK001493.1 | - | 19 - 20 | 1966 | 86 |
AK022436.1 | - | 19 - 20 | 1995 | 86 | |
AK055452.1 | - | 1 - 2 | 1183 | 86 | |
AK094022.1 | - | 10 - 11 | 2007 | 86 | |
AK097946.1 | - | 11 - 12 | 1904 | 86 | |
AL512694.1 | - | 6 - 7 | 3058 | 86 | |
BC003638.2 | - | 19 - 20 | 1988 | 86 | |
BC003666.2 | - | 19 - 20 | 1988 | 86 | |
CR457286.1 | - | 19 - 20 | 1899 | 86 |