Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCAF6
Entrez Gene ID
55827
Gene Name
DDB1 and CUL4 associated factor 6
Gene Aliases
1200006M05Rik, ARCAP, IQWD1, MSTP055, NRIP, PC326
Chromosome Location
Chr.1: 167936247 - 168075843 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017977.2 NP_001017977.1 18 - 19 2876 74
NM_001198956.1 NP_001185885.1 21 - 22 3149 74
NM_001198957.1 NP_001185886.1 20 - 21 3056 74
NM_018442.3 NP_060912.2 19 - 20 2936 74
XM_005245331.4 XP_005245388.1 20 - 21 3048 74
XM_005245332.4 XP_005245389.1 20 - 21 2919 74
XM_005245333.4 XP_005245390.1 19 - 20 2859 74
XM_017001779.1 XP_016857268.1 21 - 22 2962 74
XM_017001780.1 XP_016857269.1 19 - 20 2685 74
XM_017001781.1 XP_016857270.1 18 - 19 2675 74
GenBank mRNA AF116725.1 - 21 - 22 2890 74
AF124434.1 - 17 - 18 2367 74
AF150734.1 - 16 - 17 2236 74
AK226181.1 - 18 - 19 2727 74
AK297847.1 - 20 - 21 2916 74
AL136738.1 - 11 - 12 1521 74
AY766164.1 - 18 - 19 2616 74
BC025262.1 - 10 - 11 1539 74
BQ028917.1 - 1 - 2 74
CA310484.1 - 1 - 2 74
DQ768089.1 - 18 - 19 2540 74

View Full Product Details