The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024074.2 | NP_076979.1 | 2 - 3 | 370 | 91 |
GenBank mRNA | AK025981.1 | - | 2 - 3 | 370 | 91 |
AK292764.1 | - | 2 - 3 | 355 | 91 | |
BC001195.1 | - | 2 - 3 | 286 | 91 |