The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104.3 | NP_001095.2 | 2 - 3 | 361 | 84 |
NM_001258371.2 | NP_001245300.2 | 2 - 3 | 509 | 84 | |
NR_047663.2 | 2 - 3 | 361 | 84 | ||
GenBank mRNA | AK125851.1 | - | 2 - 3 | 895 | 84 |
AK303044.1 | - | 2 - 3 | 509 | 84 | |
AK313556.1 | - | 2 - 3 | 299 | 84 | |
BC099647.3 | - | 2 - 3 | 282 | 84 | |
BC099649.3 | - | 2 - 3 | 282 | 84 | |
DA915904.1 | - | 2 - 3 | 363 | 84 | |
M86407.1 | - | 2 - 3 | 282 | 84 |