The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145101.1 | NP_001138573.1 | 2 - 3 | 413 | 71 |
XM_011545212.2 | XP_011543514.1 | 2 - 3 | 1672 | 71 | |
XM_017018127.1 | XP_016873616.1 | 1 - 2 | 2112 | 71 | |
XM_017018128.1 | XP_016873617.1 | 2 - 3 | 369 | 71 | |
GenBank mRNA | BC157853.1 | - | 1 - 2 | 318 | 71 |
BC157854.1 | - | 1 - 2 | 318 | 71 |