Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSD4
Entrez Gene ID
23550
Gene Name
pleckstrin and Sec7 domain containing 4
Gene Aliases
EFA6B, TIC
Chromosome Location
Chr.2: 113157399 - 113203229 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012455.2 NP_036587.2 11 - 12 2407 58
XM_005263634.2 XP_005263691.1 11 - 12 2472 58
XM_006712392.2 XP_006712455.1 11 - 12 2419 58
XM_006712393.2 XP_006712456.1 11 - 12 2419 58
XM_006712394.2 XP_006712457.1 11 - 12 2419 58
GenBank mRNA AB208798.1 - 10 - 11 2846 58
AJ459781.1 - 10 - 11 2245 58
AK023421.1 - 9 - 10 1296 58
AK290045.1 - 11 - 12 2417 58
BC035307.1 - 11 - 12 2309 58
BC073151.1 - 11 - 12 2369 58
U63127.1 - 10 - 11 2287 58

View Full Product Details