Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCH7
Entrez Gene ID
64844
Gene Name
membrane associated ring-CH-type finger 7
Gene Aliases
AXO, AXOT, MARCH-VII, RNF177
Chromosome Location
Chr.2: 159712457 - 159768583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282805.1 NP_001269734.1 6 - 7 811 144
NM_001282807.1 NP_001269736.1 3 - 4 558 144
NM_022826.3 NP_073737.1 4 - 5 683 144
XM_005246773.2 XP_005246830.1 4 - 5 706 144
XM_005246774.1 XP_005246831.1 4 - 5 706 144
XM_017004718.1 XP_016860207.1 4 - 5 683 144
GenBank mRNA AK022973.1 - 4 - 5 641 144
AK222880.1 - 4 - 5 633 144
AK292836.1 - 6 - 7 779 144
AK302347.1 - 3 - 4 507 144
AK310238.1 - 3 - 4 515 144
AK316382.1 - 3 - 4 515 144
BC065014.1 - 4 - 5 607 144
BI462287.1 - - 564 145

View Full Product Details