The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318226.1 | NP_001305155.1 | 16 - 17 | 2288 | 98 |
XM_011529193.1 | XP_011527495.1 | 15 - 16 | 2226 | 98 | |
GenBank mRNA | EU718622.1 | - | 16 - 17 | 2274 | 98 |