The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005246773.2 | XP_005246830.1 | 1 - 2 | 170 | 127 |
XM_005246774.1 | XP_005246831.1 | 1 - 2 | 170 | 127 | |
XM_017004719.1 | XP_016860208.1 | 1 - 2 | 173 | 127 | |
XM_017004721.1 | XP_016860210.1 | 1 - 2 | 165 | 127 |