The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165966.1 | NP_001159438.1 | 1 - 2 | 172 | 90 |
NM_031220.3 | NP_112497.2 | 1 - 2 | 172 | 90 | |
XM_011524015.2 | XP_011522317.1 | 1 - 2 | 112 | 90 | |
XM_011524016.2 | XP_011522318.1 | 1 - 2 | 113 | 90 | |
XM_011524017.2 | XP_011522319.1 | 1 - 2 | 114 | 90 | |
GenBank mRNA | AF334586.1 | - | 1 - 2 | 169 | 90 |
BC128583.1 | - | 1 - 2 | 109 | 90 | |
BC128584.1 | - | 1 - 2 | 109 | 90 |