Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PITPNM3
Entrez Gene ID
83394
Gene Name
PITPNM family member 3
Gene Aliases
ACKR6, CORD5, NIR1, RDGBA3
Chromosome Location
Chr.17: 6451263 - 6556557 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031220.3 NP_112497.2 3 - 4 375 82
XM_011524015.2 XP_011522317.1 3 - 4 315 82
XM_011524016.2 XP_011522318.1 3 - 4 316 82
XM_011524017.2 XP_011522319.1 3 - 4 317 82
GenBank mRNA AF334586.1 - 3 - 4 372 82
AL389994.1 - 1 - 3 98 82
BC128583.1 - 3 - 4 312 82
BC128584.1 - 3 - 4 312 82

View Full Product Details