The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015136.2 | NP_055951.2 | 36 - 37 | 3939 | 65 |
XM_005264973.3 | XP_005265030.1 | 36 - 37 | 3941 | 65 | |
XM_005264974.2 | XP_005265031.1 | 36 - 37 | 3941 | 65 | |
XM_006713065.1 | XP_006713128.1 | 36 - 37 | 3941 | 65 | |
XM_017005998.1 | XP_016861487.1 | 36 - 37 | 3941 | 65 | |
XM_017005999.1 | XP_016861488.1 | 36 - 37 | 3941 | 65 | |
XM_017006000.1 | XP_016861489.1 | 36 - 37 | 3941 | 65 | |
XM_017006001.1 | XP_016861490.1 | 35 - 36 | 3845 | 65 | |
XM_017006002.1 | XP_016861491.1 | 36 - 37 | 3941 | 65 | |
XM_017006003.1 | XP_016861492.1 | 36 - 37 | 3941 | 65 | |
XM_017006004.1 | XP_016861493.1 | 36 - 37 | 3941 | 65 | |
GenBank mRNA | AB052956.1 | - | 36 - 37 | 3926 | 65 |
AJ275213.1 | - | 36 - 37 | 3895 | 65 | |
BC150250.1 | - | 36 - 37 | 3945 | 65 | |
D87433.2 | - | 36 - 37 | 3924 | 65 | |
DQ786774.1 | - | 36 - 37 | 3889 | 65 |